You Searched For: Paper
24  results were found
SearchResultCount:"24"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: Cleanroom paper is designed for use in cleanroom environments, where the prevention of chemical and particle contamination is critical.
Sustainable Packaging
Supplier: VWR Collection
Description: Formulated with coated paper, without magnesium silicate, b-paper 80 or 92 g is cut, cleaned and packed in a Class 100 (ISO 5) cleanroom.
Supplier: VWR Collection
Description: Notebook paper.
Supplier: CONTEC
Description: CONTEXTâ„¢ cleanroom documentation products have been developed to provide cleanrooms with paper and notebooks that reduce the risk of contamination.
Sale
Catalog Number: (SCOI285414989)
Supplier: SI Analytics
Description: Thermo-sensitive paper for AVS printers.
UOM: 1 * 6 items
Supplier: Thermo Scientific
Description: Toray carbon paper, TGP-H-60 can be used as a gas diffusion layer in fuel cell applications. It is also suitable for use as a catalyst backing layer.
Supplier: Texwipe
Description: Medium-weight cellulose paper saturated with synthetic latex substantially reduces particle generation associated with standard papers. These papers a...
Sale
Supplier: Texwipe
Description: TexBond® is a cleanroom bond paper with exceptional contamination control characteristics needed in critical environments. It has been especially form...
Catalog Number: (818-0132)
Supplier: Texwipe
Description: TexWrite® 22, latex-saturated cellulose cleanroom documentation sheets without organic fillers.
UOM: 1 * 2.500 items
Catalog Number: (NALG6304-9811)
Supplier: Thermo Fisher Scientific
Description: These polyolefin sheets roll or fold without cracking and stay flexible at cold temperatures. They won't rip, stretch or shrink. They can be punched f...
UOM: 1 * 50 items
Catalog Number: (TEXWTX5800)
Supplier: Texwipe
Description: TexWrite® MP 10 is a 100% synthetic substrate provides extremely low levels of particle generation along with superior usability. It is designed to us...
UOM: 1 * 500 items
Catalog Number: (PERCN5376819)
Supplier: PerkinElmer
Description: Alsiflex paper for TGA6, Pyris6 TGA, TGA4000, and STA6000 istruments.
UOM: 1 * 5 items
Catalog Number: (ORIO55PR45)
Supplier: Thermo Orion
Description: Thermal printer paper for use with pH3695-5/-7 or 55PR40/49 printers.
UOM: 1 * 5 items
Supplier: Nitritex
Description: Smooth paper surface, bagged in ISO Class 4/Class 10 cleanroom, individually lot numbered.
Catalog Number: (HPLC11650542)
Supplier: COLLECTION PREFIX HPLC
Description: Thermal paper rolls for D2500/D2000, 234 mm × 50 m/pad, Printer
UOM: 1 * 10 items
Catalog Number: (ICNA07MC1030)
Supplier: MP Biomedicals
Description: Thermal paper roll, SafTest, for Microchem I printer, 57,15 mm × 24,4 m, SafTest, Printer
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 24
no targeter for Bottom