Chloramine-T (sodium salt) trihydrate
Supplier: Roth Carl
Certificates
About this item
Synonyms: Tosylchloramide sodium trihydrate, Chlorine bleaching solution, Chloramine T trihydrate, Chloramine T sodium salt trihydrate, Sodium chloro(tosyl)amide trihydrate, N-Chloro-4-toluenesulphonamide sodium salt
CAS Number:7080-50-4
:
Formula:CH₃C₆H₄SO₂NClNa·3H₂O
MDL Number:MFCD00149066
Density:1.43 g/cm³ (20 °C)
Molecular Weight:281.69 g/mol
Melting Pt:167…170 °C
Flash Pt:192 °C (closed cup)
ADR:8 III
Merck Index:13,02085
EINECS:204-854-7
UN:3263
:GHS05
:GHS08
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications