Ruthenium standard solution, 10000 mg/L Ru in 10% hydrochloric acid (from RuCl3) ARISTAR® standard for ICP
455932MEA 545 EUR
VWRC455932M
Ruthenium standard solution, 10000 mg/L Ru in 10% hydrochloric acid (from RuCl3) ARISTAR® standard for ICP
RhCl₃.3H₂O in HCl 10%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.
For metal trace analysis in AAS or ICP
Ask for your “customized multi-element standard solution” with your exact composition using the request form
Ask for a quotation
12/31/2022