You Searched For: Drum Funnels
8  results were found
SearchResultCount:"8"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (119-0389)
Supplier: JUSTRITE MANUFACTURING CO
Description: Spill control funnel is an eco-friendly, cost-effective way to collect liquids and keep drum tops clean.
UOM: 1 * 1 items
Supplier: NEW PIG
Description: Thick, double-wall polyethylene construction resists UV rays, corrosion and most chemicals.
Catalog Number: (NEWPDRM1125-RD-NPT)
Supplier: NEW PIG
Description: Patented Burpless® designs deliver no-splash filling and one-handed access for easy regulatory compliance.
UOM: 1 * 1 items
Supplier: HIRSCHMANN
Description: These large transparent funnels, made of PP, are suitable for filling large amounts of liquid.
Supplier: BÃœRKLE
Description: Made of PP, translucent.
Supplier: VITLAB
Description: PP or HDPE, translucent.
Catalog Number: (221-1153)
Supplier: SEMADENI
Description: Funnels, made of PP, for filling containers with large quantities of liquid or powdered substances quickly.
UOM: 1 * 1 items
Supplier: asecos
Description: Drum funnel, PE, with sieve and lid, with Ø 580 mm, and filling capacity 5 l
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 8 of 8
no targeter for Bottom