You Searched For: Plastic Bottles
337  results were found
SearchResultCount:"337"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: These bottles are made with 100% virgin HDPE resin.
Supplier: VWR Collection
Description: VWR® square polycarbonate bottles are ideal for storage of media, laboratory reagents, and biological buffers in temperatures ranging from −130 to +12...
New Product
Supplier: VWR Collection
Description: VWR® square containers are ideal for storage of media, laboratory reagents, and biological buffers. Eliminate washing, depyrogenation, and autoclaving...
New Product
Supplier: VWR Collection
Description: LDPE, HDPE or PP with PP screw cap, translucent and amber.
Sale
Supplier: VWR Collection
Description: VWR® sample bottles are ideal for storage of media, laboratory reagents, and biological buffers in temperatures ranging from −70 to 60 °C. Eliminate w...
New Product
Supplier: VWR Collection
Description: PET, with or without PP screw caps.
Supplier: VWR Collection
Description: Wide neck square bottles available in HDPE or PP material.
Supplier: VWR Collection
Description: LDPE, translucent.
Supplier: VWR Collection
Description: PTFE, strong walled.
Supplier: VWR Collection
Description: Bottles are made with HDPE resin.
Sale
Supplier: VWR Collection
Description: LDPE, HDPE or PP with PP screw cap.
Sale
Supplier: VWR Collection
Description: LDPE, amber.
Supplier: VWR Collection
Description: PP, translucent.
Catalog Number: (331-0316)
Supplier: VWR Collection
Description: These bottles are made of PET and are perfect for sample collection, storage and transport.
UOM: 1 * 70 items
Supplier: VWR Collection
Description: LDPE, translucent. Excellent for sample collection, storage and transporting liquids.
Supplier: VWR Collection
Description: HDPE.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 337
no targeter for Bottom