Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
Glass Tubing
https://si.vwr.com/stibo/smallweb/std.lang.all/52/98/18945298.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"11"
  List View Easy View (new)
Sort by:
Supplier:  SCHOTT ROHRGLAS
Description:   AR-Glas®, clear.
Supplier:  SCHOTT ROHRGLAS
Description:   Borosilicate glass acc. to ISO 3585, all-purpose glass for apparatus for the chemical industry, for pipelines and laboratory.
Supplier:  SCHOTT ROHRGLAS
Description:   Borosilicate glass acc. to ISO 3585, all-purpose glass for apparatus for the chemical industry, for pipelines and laboratory.
Supplier:  SCHOTT ROHRGLAS
Description:   AR-Glas®, clear.
Supplier:  SCHOTT ROHRGLAS
Description:   Borosilicate glass acc. to ISO 3585, all-purpose glass for apparatus for the chemical industry, for pipelines and laboratory.
Supplier:  SCHOTT ROHRGLAS
Description:   Borosilicate glass tubing, varying types of inner and outer shapes.
Supplier:  BOHEMIA CRISTAL HANDELSGES.MBH
Description:   Borosilicate glass, SIMAX.
Catalog Number: (HECH42840001)
Supplier:  GLASWARENFABRIK KARL HECHT
Description:   AR-Glass®, clear.
UOM:  1 * 1 kg
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 8  of 11
  1  2  Next 
A new look and a renewed commitment
Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.
Learn More Start Shopping