You Searched For: Butyrometers
13  results were found
SearchResultCount:"13"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: GERBER FUNKE DR N.
Description: According to Teichert`s method.
Catalog Number: (614-0097)
Supplier: GERBER FUNKE DR N.
Description: According to Sichler`s method, with rounded scale.
UOM: 1 * 1 items
Supplier: GERBER FUNKE DR N.
Description: According to Koehler´s measuring method.
Sale
Catalog Number: (614-0105)
Supplier: GERBER FUNKE DR N.
Description: Measuring method, for ice cream.
UOM: 1 * 1 items
Supplier: GERBER FUNKE DR N.
Description: According to Kehe`s method.
Supplier: GERBER FUNKE DR N.
Description: For drinking vat milk, frosted rear scale wall, error tolerance 0,025%.
Catalog Number: (614-0132)
Supplier: GERBER FUNKE DR N.
Description: According to Roeder´s weighing method.
UOM: 1 * 10 items
Supplier: GERBER FUNKE DR N.
Description: Borosilicate glass.
Catalog Number: (614-0135)
Supplier: GERBER FUNKE DR N.
Description: According to Roeder´s weighing method.
UOM: 1 * 10 items
Catalog Number: (614-0136)
Supplier: GERBER FUNKE DR N.
Description: For determining free fat content in milk and cream.
UOM: 1 * 1 items
Supplier: GERBER FUNKE DR N.
Description: According to Roeder´s weighing method.
Supplier: GERBER FUNKE DR N.
Description: According to Roeder´s weighing method.
Supplier: Gerber Instruments AG
Description: DURAN®, borosilicate glass.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 13 of 13
no targeter for Bottom