You Searched For: Drawer Organisation Trays
9  results were found
SearchResultCount:"9"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (631-0727)
Supplier: VWR Collection
Description: PS.
UOM: 1 * 1 items
Supplier: Metro International
Description: The tote box provide safe and efficient storage of items in bulk, or in compartmentalised fashion with the use of optional dividers and covers.
P...
Supplier: Metro International
Description: Divider tote boxes made of polypropylene base resins offer safe and efficient storage of items in bulk, or in compartmentalised fashion with the use o...
Supplier: Bel-Art Products, a Part of SP
Description: Drawer organiser, made of white polystyrene.
Catalog Number: (NALG5832-0001)
Supplier: Thermo Fisher Scientific
Description: Acrylic, clear.
UOM: 1 * 1 items
Supplier: KARTELL
Description: PVC, white.
Supplier: Brand
Description: PVC, white.
Catalog Number: (WALD81012711)
Supplier: WALDNER LABOREINRICHTUNG
Description: Drawer trays for fully equipping drawers and for underbench units with a depth of 1200 mm.
UOM: 1 * 1 items
Supplier: SEMADENI
Description: Made of PS, grey.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 9 of 9
no targeter for Bottom