Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
    • In Stock (27)
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
Joint Sleeves
https://si.vwr.com/stibo/smallweb/std.lang.all/95/91/18709591.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"16"
  List View Grid View Easy View (new)
Sort by:
Supplier:  VWR Collection
Description:   PTFE, 0,05 mm thick.
Supplier:  VWR Collection
Description:   PTFE, with grip.
Supplier:  Bohlender
Description:   Spherical ground joint sleeves, made of PTFE, with gripping rings and sealing rings on the outside; seal suitable for high vacuums. Robust design for ...
Supplier:  COWIE
Description:   Made of PTFE, grease-free, high vacuum full length rigid sleeves for standard glass joints, with substantial shoulder for ease of handling and repeate...
Supplier:  Bohlender
Description:   PTFE.
Supplier:  Bohlender
Description:   Reusable sleevesmade of PTFE, with gripping ring and sealing rings on the outside.
Supplier:  COWIE
Description:   Thin wall joint sleeves, made of PFTE, are inserted between standard taper ground glass joints to eliminate the use of grease and prevent sticking.
Catalog Number: (BOHLH981-14)
Supplier:  Bohlender
Description:   Ground joint reducing set, PTFE, consisting of reducing rings graduated for bridging different cone and socket sizes.
UOM:  1 * 1 SET
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 8  of 16
  1  2  Next