You Searched For: Gas Cartridges
7  results were found
SearchResultCount:"7"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (451-2201)
Supplier: USBECK Laborgeräte
Description: Heater, Gas cartridges, 400 ml, Butane/propane
UOM: 1 * 1 items
MSDS
Supplier: CAMPING GAZ INTERNATIONAL
Description: Heater, Gas cartridges, CV 300 Plus, 240 g, Butane/propane
Catalog Number: (CAMP2000033971)
Supplier: CAMPING GAZ INTERNATIONAL
Description: Campingaz® gas cartridge CP250 with safety valve.
UOM: 1 * 1 items
Catalog Number: (CAMP2182099)
Supplier: CAMPING GAZ INTERNATIONAL
Description: Campingaz® gas cartridge CG1750 with safety valve.
UOM: 1 * 12 items
Catalog Number: (451-4264)
Supplier: FRANKEA GMBH
Description: Universal butane gas, for gas lighters.
UOM: 1 * 1 items
Catalog Number: (HECH41396020)
Supplier: GLASWARENFABRIK KARL HECHT
Description: Heater, Gas cartridges, 190 g, Butane
UOM: 1 * 190 g
MSDS
Catalog Number: (451-0047)
Supplier: R-L ENTERPRISES LTD
Description: Heater, Gas cartridges, 220 g, Butane/propane
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 7 of 7
no targeter for Bottom