Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
    • In Stock (32)
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "ice-pans"
https://si.vwr.com/stibo/smallweb/std.lang.all/94/22/18989422.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"8"
  List View Grid View Easy View (new)
Sort by:
Supplier:  VWR Collection
Description:   Polyurethane foam.
Supplier:  BIOCISION, LLC
Description:   Ethylene vinyl acetate (EVA) foam
Supplier:  Corning Life Science
Description:   Ethylene vinyl acetate (EVA) foam
Supplier:  Bel-Art
Description:   Ice pan, Mini, with lid, Magic Touch™, Urethane, Capacity: 1 l, Colour: Blue, W×D×H: 183×183×135 mm
Supplier:  Bel-Art
Description:   These pans are ideal for ice and water baths as well as general organization and transport of samples.
Supplier:  Bel-Art
Description:   Coloured expanded polymer
Supplier:  KOEHLER TECHNISCHE PRODUKTEN
Description:   Extremely good isolated ice bath made of double-walled polystyrene. The hollow is foamed in the inside for a better temperature isolation. Due to it a...
Catalog Number: (478-1255)
Supplier:  NEOLAB
Description:   Extremely good isolated ice bath made of double-walled polystyrene. The hollow is foamed in the inside for a better temperature isolation. Due to it a...
UOM:  1 * 1 items
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 8  of 8