Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
    • In Stock (10)
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
Buckets, Metal
https://si.vwr.com/stibo/smallweb/std.lang.all/96/90/18709690.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"9"
  List View Easy View (new)
Sort by:
Supplier:  BOCHEM
Description:   Buckets, stainless steel 18/10.
Supplier:  RSG
Description:   Stainless steel, rust-proof, polished finish.
Supplier:  RSG
Description:   Lid for bucket, Stainless steel
Supplier:  BOCHEM
Description:   Stainless steel 18/10.
Supplier:  BOCHEM
Description:   Lid, Ø 270 mm, For: Jug, 15 l capacity
Supplier:  Texwipe
Description:   Stainless steel, Type 304.
Catalog Number: (USBE4500)
Supplier:  USBECK Laborgeräte
Description:   Stainless steel.
UOM:  1 * 1 items
Supplier:  COLDSTREAM ENGENEERING
Description:   Stainless steel, 304 grade.
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 8  of 9
  1  2  Next 
A new look and a renewed commitment
Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.
Learn More Start Shopping