You Searched For: Blue Light Transilluminators
5  results were found
SearchResultCount:"5"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: Blue light transilluminators are often used as an alternative to UV transilluminators when users wish to use ‘safe dyes’ instead of ethidium bromide. ...
Catalog Number: (SYNOGX-ULTRABRIGHT)
Supplier: Syngene (a division of the Synoptics group)
Description: Blue light transilluminators are often used as an alternative to UV transilluminators when users wish to use ‘safe dyes’ instead of ethidium bromide. ...
UOM: 1 * 1 items
Catalog Number: (SYNOGEULTRASLIMLED)
Supplier: Syngene (a division of the Synoptics group)
Description: UltraSlim-LED transilluminators for superior gel illumination.
UOM: 1 * 1 items
Supplier: UVP ULTRA VIOLET PRODUCTS LTD
Description: Visi-Blue™ transilluminators are designed for use with SYBR® Gold, SYBR® Green, GFP and other 460 to 470 nm blue excitation samples.
Catalog Number: (733-2005)
Supplier: EDVOTEK EUROPE LTD
Description: The TruBlu™ blue light transilluminator is ideal for viewing DNA gels stained with SYBR® Safe, thus eliminating the need for UV light or ethidium brom...
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 5 of 5
no targeter for Bottom