You Searched For: UV Transilluminators
7  results were found
SearchResultCount:"7"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: All VWR® transilluminators slide out for easy access to provide very even illumination. Those for use with the VWR® Imager system also have variable i...
Catalog Number: (730-1488)
Supplier: VWR Collection
Description: The VWR® benchtop transilluminator for stand alone imaging systems is designed for ease of use.
UOM: 1 * 1 items
Supplier: Syngene (a division of the Synoptics group)
Description: UV Transilluminator, Syngene, GX-2530L-E, Wavelength: 365 nm, Filter size: 250×230 mm
Catalog Number: (733-2310)
Supplier: Syngene (a division of the Synoptics group)
Description: UV Transilluminator, Syngene, GE-2020M-E, Wavelength: 302 nm, Filter size: 200×200 mm
UOM: 1 * 1 items
Supplier: UVP ULTRA VIOLET PRODUCTS LTD
Description: These High Performance UV transilluminators, with four 25 W tubes, provide 100 W of high intensity and uniform UV illumination for high fluorescence a...
Supplier: UVP ULTRA VIOLET PRODUCTS LTD
Description: All Benchtop transilluminators are compact, lightweight and offer an economical alternative to the High Performance series transilluminators. With the...
Catalog Number: (UVPA38-0325-01)
Supplier: UVP ULTRA VIOLET PRODUCTS LTD
Description: UV Transilluminator
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 7 of 7
no targeter for Bottom