You Searched For: Baths with Integral Circulators
6  results were found
SearchResultCount:"6"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: Rozsdamentes acél termosztátok Advanced Programmable vagy Advanced Digital hőmérsékletszabályozóval. Mindkettő könnyen navigálható a nagyméretű, kijel...
Supplier: JULABO GmbH
Description: The DYNEOâ„¢ DD heating circulators are available in several bath combinations, with varying bath sizes. DYNEO is designed for internal as well as exter...
Supplier: JULABO GmbH
Description: The MAGIO™ series of heating circulators operate with a working temperature range of 20 to +300 °C and heating capacity of up to 3 kW. With their powe...
Supplier: JULABO GmbH
Description: The powerful and robust CORIOâ„¢ laboratory circulators provide the exact temperature, absolute precision, and a wide working temperature range. The COR...
Catalog Number: (461-1219)
Supplier: LAUDA
Description: The Proline Edition X range of heating thermostats feature a wide temperature range, high cooling and heating power and a multi level pump. The self-e...
UOM: 1 * 1 items
Supplier: HUBER
Description: The low bath volume means these circulators are ideal for controlling the temperature of small external applications, or for direct temperature contro...
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 6 of 6
no targeter for Bottom