Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
    • In Stock (20)
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "overhead-stirrers-electronic"
SearchPresentationType-VERTICAL
SearchResultCount:"11"
  List View Grid View Easy View (new)
Sort by:
Supplier:  VWR Collection
Description:   This quiet, economical overhead stirrer is suitable for simple laboratory mixing, applications such as processing media with substantial temporary vis...
Supplier:  VWR Collection
Description:   Laboratory stirrers which are ideal for simple stirring tasks, suitable for quantities up to 25 litres (VOS 40 digital), or for intensive stirring tas...
Supplier:  VWR Collection
Description:   Lightweight, ergonomic overhead stirrer which is ideal for high viscosity mixing, large batch mixing or stirring products with medium viscosity. Micro...
Supplier:  IKA
Description:   Extremely powerful laboratory stirrers for highly viscous applications and intensive mixing of quantities up to 100 l (H₂O). Units automatically adjus...
Supplier:  IKA
Description:   Powerful laboratory stirrers for quantities up to 40 or 100 l (H₂O). Units automatically adjust the speed through microprocessor controlled technology...
Supplier:  IKA
Description:   Laboratory stirrers designed for simple stirring tasks for quantities up to 15 or 25 l (EUROSTAR 20 digital and EUROSTAR 40 digital), or for intensive...
Supplier:  VELP SCIENTIFIC
Description:   Range of overhead stirrers with electronic adjustment that are equipped with advanced safety systems.
Supplier:  HEIDOLPH
Description:   Powerful electronic stirrers that feature a smooth start operation which prevents spills and splashing media. The speed ramps up slowly until set spee...
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 8  of 11
  1  2  Next