Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
    • In Stock (33)
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "laboratory-shoe-and-boot-covers"
SearchPresentationType-VERTICAL
SearchResultCount:"27"
  List View Grid View Easy View (new)
Sort by:
Supplier:  VWR Collection
Description:   Shoe covers made from 65% Low Density PE (LDPE) and 35% Linear Low Density PE (LLDPE).
Supplier:  VWR Collection
Description:   Shoe covers made from non woven PP. With latex pattern.
UOM:  1 * 100 items
Supplier:  VWR Collection
Description:   These non woven shoe covers are made of PP with a skid-resistant CPE coating on the sole.
Supplier:  VWR Collection
Description:   Basic shoe covers manufactured from a low-density chlorinated polyethylene fabric, which is designed to protect against dirt, grime, and liquid splash...
Catalog Number: (113-8280)
Supplier:  VWR Collection
Description:   Shoe covers made from non woven PP.
UOM:  1 * 100 items
Supplier:  VWR Collection
Description:   Non woven PP with a CPE laminated sole.
Catalog Number: (HOIT110176)
Supplier:  VWR Collection
Description:   These disposable shoe covers are made of CPE.
UOM:  1 * 2.000 items
Catalog Number: (113-8287)
Supplier:  VWR Collection
Description:   Shoe covers made from non woven PP.
UOM:  1 * 300 items
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 8  of 27
  1  2  3  4  Next