Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
    • In Stock (99)
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "Cleanroom Overalls"
SearchPresentationType-VERTICAL
SearchResultCount:"22"
  List View Grid View Easy View (new)
Sort by:
Supplier:  VWR Collection
Description:   The VWR Basic SPP overalls are manufactured from a strong, lightweight, and breathable spunbonded polypropylene (SPP) fabric, which protects against d...
Supplier:  VWR Collection
Description:   VWR Advanced SF overalls are manufactured from spunbonded polypropylene fabric coated with breathable polyethylene film.
Supplier:  VWR Collection
Description:   These cleanroom overalls are manufactured from a latex-free, multilayer spunbonded-meltblown-spunbonded (SMS) fabric that offers high tensile strength...
Supplier:  VWR Collection
Description:   VWR Maximum SF overalls are the cleanest and most durable within the entire VWR clothing line. Made from spunbonded polypropylene fabric (SPP) coated ...
Supplier:  VWR Collection
Description:   This single-use garment set includes overall, hood, boot covers and sleeve protectors, all made from breathable SMS fabric.
Supplier:  VWR Collection
Description:   These sterile single-use overalls are made of breathable SMS fabric. They are ideal for use in cleanroom production and research.
Supplier:  VWR Collection
Description:   Disposable, anti-static overall made of a microporous film with a nylon scrim between film and substrate that gives the material additional strength.
Supplier:  VWR Collection
Description:   These overalls are made from Protect Xtra fabric which is a further development of the Protect fabric consisting of 100% polyester weave with conducti...
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 8  of 22
  1  2  3  Next