You Searched For: Respirator Filters
30  results were found
SearchResultCount:"30"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: 3M
Description: Respirator Filter, 501 Filter retainer, For: 3Mâ„¢ 5000, 6000 Series
Sale
Supplier: 3M
Description: 3Mâ„¢ Secure Clickâ„¢ D3000 Series Particulate Filters feature a new dual flow design for easier breathing.
Sale
Supplier: 3M
Description: 3Mâ„¢ Secure Clickâ„¢ D7000 series particulate filters are designed for use with Secure Clickâ„¢ 800 series respirators, Secure Clickâ„¢ D8000 series filters ...
Catalog Number: (MMMA7100260645)
Supplier: 3M
Description: For effective protection against a range of particulates, gases and vapours or combination hazards, use 3Mâ„¢ Filters DT-Series. Suitable for use with c...
UOM: 1 * 1 items
New Product
Supplier: 3M
Description: These mask filters are light, comfortable and provide reliable protection from particulates. When using with 3Mâ„¢ 6000 series masks, a 3Mâ„¢ 502 adapter ...
Supplier: 3M
Description: The 3M 6000 Series Gas, Vapor, and Particulate Combination Filters have a bayonet-style connection that fits any half and full-face 3M reusable masks....
Supplier: 3M
Description: 3Mâ„¢ Secure Clickâ„¢ cartridges feature a new dual-flow design to allow for easier breathing.
Sale
Supplier: 3M
Description: These filters provide protection against dusts, mists and other particulate hazards when used with the 3Mâ„¢ Versaflo Powered Air Respirator, TR-300 Ser...
Catalog Number: (MMMA7100260622)
Supplier: 3M
Description: For effective protection against a range of particulates, gases, and vapours, or combination hazards, use 3Mâ„¢ filters DT series. Suitable for use with...
UOM: 1 * 10 items
New Product
Catalog Number: (111-2566)
Supplier: 3M
Description: Used to hold 3M Secure Clickâ„¢ particulate filters D7N11 or D7P71 in place on Secure Clickâ„¢ D8000 series cartridges.
UOM: 1 * 10 items
Catalog Number: (111-2145)
Supplier: SCOTT SAFETY
Description: Combining low weight and low breathing resistance Pro2000 filters are manufactured using superior performance media, giving extended adsorption capaci...
UOM: 1 * 20 items
Supplier: Moldex
Description: EasyLock® filters protect against gas, vapour and dust. Suitable for series 7000 and 9000 masks.
Supplier: Dräger
Description: Range with a Dräger-specific bayonet connection that fits X-plore® 3300/3500 half masks and X-plore® 5500 full face masks.
Supplier: Dräger
Description: This series of filters with Dräger specific thread connection Rd90 (91 mm diameter) was designed for use in half masks. Suitable for Dräger half masks...
Supplier: Dräger
Description: The filters of the Dräger X-plore® Rd90 series protect from hazardous gases and vapors, have a long life and can be used for many different masks.
Supplier: Dräger
Description: The X-plore® Rd40 filter with aluminium housing is suitable for half and full face masks with connectors conforming to EN 148-1.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 30
no targeter for Bottom