You Searched For: Floor Mats
56  results were found
SearchResultCount:"56"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: 30 or 60 layer adhesive entrance mats manufactured from low density polyethylene. These mats control contamination by removing debris from the soles o...
Supplier: VWR Collection
Description: These mats offer the comfort of a polyurethane mat which is suitable for ISO Class 5 cleanroom (Class 100) environments. It is either available with a...
Supplier: VWR Collection
Description: These mats are made of low density polyurethane providing an ergonomic, anti-fatigue profile. Bubble mats are designed for dry, non critical environme...
Supplier: VWR Collection
Description: These frames are designed to be used with PureStep mats to create a system, that is ideal for any critical area, and does not allow direct adhesion to...
Supplier: VWR Collection
Description: Permanent mats are installed to provide the highest level of cleanliness. The life of mats strongly depends on the volume, and degree of cleanliness o...
Supplier: VWR Collection
Description: B'mat decontamination flooring uses a patented polymer to attract and retain over 98% of foot- and wheel-borne particles. Particles are naturally attr...
Catalog Number: (115-1060)
Supplier: VWR Collection
Description: Anti-fatigue mats, VWR®, For cleanroom use: +, L×W: 600×900 mm
UOM: 1 * 1 items
Catalog Number: (590-0346)
Supplier: Bohlender
Description: Mat made of static dissipative material with permanently mounted grounding cable including clip for earthing connection.
UOM: 1 * 1 items
Supplier: 3M
Description: 4300 contamination control mats consist of 40 layers of adhesive-coated clear polyethylene sheets attached to a vinyl backing. For use at cleanroom en...
Sale
Supplier: JUSTRITE MANUFACTURING CO
Description: Anti-Stat P.O.P.â„¢ is a vinyl work surface that dissipates static from conductive items placed on it.
Supplier: Texwipe
Description: Adhesive coated contamination control mats capture particulates from foot traffic and equipment wheels, providing the most effective dirt and dust rem...
Supplier: 3M
Description: The Nomadâ„¢ Aqua Textile Drop Down Mat 65 is a high-performance entrance mat designed for medium traffic applications.
Catalog Number: (115-2586)
Supplier: NEW PIG
Description: Adhesive-backed mat runner lets you create custom-sized restroom mats for urinals, sinks and other wet spots.
UOM: 1 * 1 Roll
Supplier: 3M
Description: The Nomadâ„¢ Aqua Textile Drop Down Mat 85 is a high-performance entrance mat with a phthalate-free, slip-resistant PVC backing for high traffic applica...
Supplier: 3M
Description: The Nomadâ„¢ Aqua Textile Drop Down Mat 45 is a high-performance entrance mat backed with slip-resistant PVC for low traffic applications.
Supplier: 3M
Description: The Nomadâ„¢ Terra Heavy Traffic Entrance Matting 8100 is unbacked scraper matting suitable for use indoors, outdoors, or in recessed wells with high tr...
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 56
no targeter for Bottom