Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "insulating-tapes"
https://si-bot.vwr.com/stibo/smallweb/std.lang.all/94/43/18989443.jpg
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
SearchResultCount:"21"
  List View Grid View Easy View (new)
Sort by:
Catalog Number: (332362S)
Supplier:  VWR Chemicals
Description:   Colour: white
UOM:  1 * 1 Roll
Catalog Number: (BRDY266081)
Supplier:  BRADY & CO LTD, W. H.
Description:   Protect printed messages with this clear overlaminating tape.
UOM:  1 * 1 Roll
Supplier:  Bohlender
Description:   Deformable, virginal PTFE flat tape with expanded fibre structure. For making customized gaskets “on-site”. FDA conform.
Catalog Number: (291-0041)
Supplier:  Bohlender
Description:   PTFE tape for sealing joints and screw threads. Thickness: 0,1 mm
UOM:  1 * 1 Roll
Catalog Number: (BURK0006-4514)
Supplier:  BÜRKLE
Description:   TAPE SEALING F/THREADS PTFE 12MX12MM 1 * 1 items
UOM:  1 * 1 items
Supplier:  3M
Description:   Scotch™ Performance Flatback Tape is used in holding, splicing or masking applications where high adhesion strength, high temperature capability and h...
Supplier:  TQC
Description:   This tape consists of 23 microns polyester films backing for improved moisture resistance, weight and space savings. These tapes are ideal to perform ...
Catalog Number: (819-1400)
Supplier:  3M
Description:   Aluminium/acrylic.
UOM:  1 * 1 Roll
Discontinued
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 8  of 21
  1  2  3  Next