You Searched For: Rubber Stoppers
39  results were found
SearchResultCount:"39"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: Saint-Gobain
Description: Conical natural grey-blue rubber stoppers without holes.
Supplier: Simport Scientific
Description: Made of thermoplastic elastomer these disposable closures produce a firm, leak-resistant seal for glass and plastic tubes. They will protect samples f...
Supplier: Saint-Gobain
Description: The white stoppers made of natural rubber guarantee absolute impermeability, even when the edges of the tubes or glass, metal or plastic containers to...
Supplier: Saint-Gobain
Description: Conical natural grey-blue rubber stoppers without holes.
Supplier: DWK Life Sciences
Description: Rubber stopper, WHEATON®
Supplier: Saint-Gobain
Description: Natural rubber, grey-blue.
Supplier: DWK Life Sciences
Description: This bromobutyl rubber closure provides a gas-tight seal for all GL 25 and GL 45 laboratory bottles with sizes from 10 to 20000 ml.
Supplier: KARTELL
Description: Conical red rubber stoppers with one or two holes.
Supplier: DEUTSCH & NEUMANN
Description: Rubber stopper, Red, Hole Ø: 6,0 mm, Ø bottom: 12,0 mm, Ø top: 19,0 mm, Height: 18,0 mm
Supplier: DEUTSCH & NEUMANN
Description: Rubber stopper, Red, No. of holes: 2, Hole Ø: 5,0 mm, Ø bottom: 23,0 mm, Ø top: 27,0 mm, Height: 30,0 mm
Supplier: DEUTSCH & NEUMANN
Description: Rubber stopper, Black, Ø bottom: 41,0 mm, Ø top: 49,0 mm, Height: 40,0 mm
Supplier: DEUTSCH & NEUMANN
Description: Natural rubber, grey.
Supplier: DEUTSCH & NEUMANN
Description: Stopper, chloroprene rubber, Reddish brown, No. of holes: 1, Hole Ø: 5,0 mm, Ø bottom: 21,0 mm, Ø top: 27,0 mm, Height: 30,0 mm
Supplier: KARTELL
Description: Natural red rubber with elastic, tight fitting design.
Supplier: DEUTSCH & NEUMANN
Description: Conical natural grey rubber stoppers with one hole.
Catalog Number: (DENE2100814)
Supplier: DEUTSCH & NEUMANN
Description: Rubber stopper, Red, Hole Ø: 8,0 mm, Ø bottom: 14,0 mm, Ø top: 20,0 mm, Height: 32,0 mm
UOM: 1 * 100 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 39
no targeter for Bottom