You Searched For: Automatic Transmitance Colorimeters
7  results were found
SearchResultCount:"7"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: Lovibond Tintometer
Description: A series of automatic spectrophotometric colorimeters for objective colour analysis of edible oils, chemicals, petrochemicals, fuels and other transpa...
Supplier: Lovibond Tintometer
Description: Accessory for colorimeter, Automatic, transmittance, ICUMSA colour upgrade (420 nm, 560 nm, 710 nm)
Supplier: Lovibond Tintometer
Description: Two spectrophotometric colorimeters that each include several of the popular scales that meet the requirements of objective colour analysis of oils, c...
Supplier: Lovibond Tintometer
Description: The PFX995 spectrophotometers are top of the range, versatile instruments that offer comprehensive colour data requirements for either t...
Supplier: Lovibond Tintometer
Description: A series of spectrophotometric colorimeters for colour grading of optically clear samples. Each version of the PFXi 195 includes a selection of...
Supplier: Lovibond Tintometer
Description: The Model F is a visual colorimeter designed to optimise the use of Lovibond® glass filters in order to express colourof the sample in terms of Lovibo...
Catalog Number: (TINT107103)
Supplier: Lovibond Tintometer
Description: The AF710-3 is a visual instrument used for colour grading of oils and fats.
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 7 of 7
no targeter for Bottom