You Searched For: Transport Swabs
19  results were found
SearchResultCount:"19"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: Collection and transport swabs for bacterial specimens with applicators and transport media.
Sale
Catalog Number: (710-0442)
Supplier: VWR Collection
Description: Transport swab with plastic shaft, synthetic tip, and Amies agar gel.
UOM: 1 * 50 items
Supplier: KIMA SAS DI CHIARIN RENZO & C
Description: Swabs in test tube, Amies gel with charcoal, Aluminium, Ø: 12 mm, 140 mm
Supplier: COPAN
Description: A range of collection and transport devices for bacterial specimens.
Supplier: THERMO FISHER DIAGNOSTICS GMBH
Description: Effective transportation of throat, vaginal, wound and skin swabs conforming to highest medical device classification - Class II a surgically invasive...
Supplier: COPAN
Description: Liquid transport medium is contained in a sponge carrier located in the bottom of the tube.
Catalog Number: (COPA414C)
Supplier: COPAN
Description: The M40 Transystem line is the latest generation of traditional bacteriology transport swabs. It was designed to provide the ultimate performance capa...
UOM: 1 * 50 items
Supplier: DELTALAB
Description: Snappable transport swabs in a tube.
Supplier: DELTALAB
Description: The glycerophosphate has been substituted by inorganic phosphate since glycerophosphate is a metabolite for some bacteria, so they could grow and disg...
Supplier: DELTALAB
Description: The modified Stuart medium allows the conservation and transport of a large number of pathological microorganisms
Catalog Number: (DELT300299)
Supplier: DELTALAB
Description: Transport swab suitable for the detection of Chlamydia.
UOM: 1 * 600 items
Supplier: BALIS
Description: Swabtube, with 10 ml peptone pysiolog. saline solution, VWR®
Supplier: MEDICAL WIRE & EQUIPMENT
Description: Each transport tube contains Amies medium preferred because the inorganic buffer ensures maintenance without overgrowth.
Supplier: MEDICAL WIRE & EQUIPMENT
Description: Transwab with a narrow shaft and bud for nasopharyngeal or urethral sampling.
Supplier: HERENZ, HEINZ GMBH
Description: Microbiology transport sets sterilised by radiation.
Catalog Number: (LPIT118898)
Supplier: LP ITALIANA SPA
Description: Swabs with transport medium.
UOM: 1 * 250 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 19
no targeter for Bottom