You Searched For: Reusable Earplugs
22  results were found
SearchResultCount:"22"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (111-1629)
Supplier: 3M
Description: Corded earplugs made from a patented polymer formulation, the comfortable triple flanged tip of these earplugs provides an excellent attenuation of 35...
UOM: 1 * 50 PAIR
Supplier: 3M
Description: The unique, one size E-A-Râ„¢ Tracerâ„¢ earplugs fit most ear canals comfortably and provide high levels of attenuation with the added benefit of incorpor...
Supplier: 3M
Description: E-A-R™ Express™ Reusable Earplugs are recognisable yellow and blue earplugs with a foam tip that’s shaped and sized for a comfortable fit. They can re...
Supplier: 3M
Description: E-A-Râ„¢ Flexible Fit HA earplugs have a patented earplug foam formulation which makes them fully washable and reusable. The soft foam tip and flexible ...
Supplier: 3M
Description: Earplugs manufactured from a soft elastic material to ensure a secure and comfortable fit.
Catalog Number: (MMMAUF01015)
Supplier: 3M
Description: These pre-moulded earplugs made from soft TPE use unique filter technology to take the edge off low levels of harmful noise whilst allowing normal con...
UOM: 1 * 200 PAIR
Supplier: 3M
Description: These earplugs are made from flexible materials that are already pre-formed to ensure optimum fit.
Catalog Number: (MMMAUF01000S)
Supplier: 3M
Description: The 3Mâ„¢ E-A-Râ„¢ UltraFitâ„¢ Earplugs are reusable and designed for insertion into the ear canal to help reduce exposure to hazardous levels of noise up t...
UOM: 1 * 200 PAIR
Catalog Number: (MMMAEEP-100EU)
Supplier: 3M
Description: Peltor™ Electronic Earplugs have an intuitive one-button operation that turns the earplugs on, off, and selects volume settings. They’re powered by Li...
UOM: 1 * 1 PAIR
Catalog Number: (MMMAPN01006)
Supplier: 3M
Description: 3Mâ„¢ Tri-Flangeâ„¢ Earplugs are innovative, comfortable hearing protectors with soft, flexible flanges that adapt to the shape of ears.
UOM: 1 * 400 PAIR
Catalog Number: (MMMAUF01012)
Supplier: 3M
Description: These pre-moulded earplugs are suitable for moderate noise levels (83 to 93 dB(A)). Correctly matched to the noise level they allow conversation and b...
UOM: 1 * 50 PAIR
Supplier: Moldex
Description: These earplugs provide serious protection for use in most noisy environments.
Sale
Catalog Number: (111-0720)
Supplier: Moldex
Description: Corded earplugs made from soft TPE material which is gentle on the skin. The flexible design and the solid handle ensures easy insertion and removal o...
UOM: 1 * 50 PAIR
Catalog Number: (MOLD644101)
Supplier: Moldex
Description: Twisters® earplugs have a soft foam pod for a comfortable, snug seal.
UOM: 1 * 80 PAIR
Catalog Number: (111-1163)
Supplier: Moldex
Description: These reusable earplugs provide protection against lower noise levels. They are designed to keep the user in touch with the environment.
UOM: 1 * 50 PAIR
Catalog Number: (UVEX2111.239)
Supplier: UVEX
Description: Earplugs with cord are conveniently at hand, whenever required. Suitable for use in dirty working environments.
UOM: 1 * 50 PAIR
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 22
no targeter for Bottom