You Searched For: Noise Blocking Earmuffs
23  results were found
SearchResultCount:"23"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (MMMAMRX214AWS6)
Supplier: 3M
Description: The 3M™ PELTOR™ WS™ ALERT™ X headset is a Bluetooth® headset with noise-cancelling boom microphone that connects to a mobile phone for hands-free call...
UOM: 1 * 1 items
Supplier: 3M
Description: These earmuffs combine excellent protection with ultimate comfort. They are available as headband and helmet-mounted versions, offering a wide range o...
Supplier: 3M
Description: These earmuffs have been developed for use in extremely high noise environments, particularly those dominated by low frequency sounds. The protection ...
Supplier: 3M
Description: The lightweight Peltorâ„¢ Optimeâ„¢ I earmuffs provide a moderate level of attenuation that meets the needs of the majority of industrial applications.
Supplier: 3M
Description: Optimeâ„¢ II earmuffs are available in various designs for different requirements: With foldable headband, neckband or as a helmet attachment.
Supplier: JSP LTD
Description: The Sonis® ear defender range has been designed with a colour coded system for simple selection, and peaks at an unbeatable 37 SNR.
Supplier: 3M
Description: ProTacâ„¢ III hearing protector helps protect against harmful noise, while allowing ambient sounds to be heard at a level below 82 dB. It increases the ...
Supplier: 3M
Description: Rechargeable battery pack for use with specific 3Mâ„¢ Peltorâ„¢ headsets.
Supplier: 3M
Description: X series earmuffs are 3M’s latest advancement in hearing conservation. New technologies in comfort, design and protection all come together in this gr...
New Product
Catalog Number: (MMMA393-1100)
Supplier: 3M
Description: Fit testing is key for effective wear and hearing conservation. The 3Mâ„¢ E-A-Rfitâ„¢ Dual Ear Validation System accurately quantifies personal attenuatio...
UOM: 1 * 1 KIT
Catalog Number: (MMMAX5P5E)
Supplier: 3M
Description: 3Mâ„¢ PELTORâ„¢ X5 Earmuffs are innovative hearing protectors that offer exceptional performance, without the need for double protection. Offering the sam...
UOM: 1 * 1 items
Catalog Number: (MMMAX1P5E-OR)
Supplier: 3M
Description: 3Mâ„¢ PELTORâ„¢ X1 Earmuffs are lightweight hearing protectors with comfortable, soft ear cushions. They feature an electrically insulated, twin headband ...
UOM: 1 * 10 items
Catalog Number: (PRTWPW43BKR)
Supplier: Portwest
Description: Comfort and robust ear muffLightweight for all day wearPivoting pin cups for better fitPadded headband for comfortHigh attenuation rate.
UOM: 1 * 1 items
Supplier: Portwest
Description: Classic ear protectors made from robust ABS.
Catalog Number: (PRTWPW47RER)
Supplier: Portwest
Description: These helmet-mounted ear protectors made from ABS feature an excellent attenuation level.
UOM: 1 * 1 items
Catalog Number: (MOLD6100)
Supplier: Moldex
Description: M1 Premium foldable earmuff with exclusive metallic colour and SoftCoat® covering that helps muffle transmitted noise.
UOM: 1 * 1 items
Safer Human and Environmental Health
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 23
no targeter for Bottom