Ultrapure water system, Milli-Q® Reference
Supplier: Merck
Milli-Q®
MILLZ00QSVCFREA 8760 EUR
MILLZ00QSVCFR 171-0237 171-0238 171-0235 171-0236 171-0234 171-0242 171-0243 171-0240 171-0296 171-0241
Ultrapure water system, Milli-Q® Reference
Water Purification Systems Pure and Ultra Pure Water Systems
Milli-Q® Reference system delivers ultrapure (Type I) water customised to fit a broad range of research areas while conserving laboratory resources such as time and space. It incorporates a new purification strategy combining initial purification of Type II feed water with individualised final purification. Depending on their application, scientists can install a variety of available polishing cartridges to remove specific contaminants at the point of delivery. A graphic screen clearly displays water quality and system status. Combining deionisation, UV technology and final polishing, the Milli-Q® Reference system produces Type I ultrapure water (18,2 MOhm.cm resistivity @ 25 °C, ≤5 ppb total organic carbon (TOC)) on demand. The innovative purification strategy allows the use of different consumables at the final polishing step to target the contaminants that are critical to different applications. Produces Type I reagent grade water from pre-treated feed water.
  • Volumetric function saves time - automatic delivery of the water volumes that are needed
  • The low flow function allows easy preparation of solutions
  • Graphic screen clearly displays water quality and system status information in a choice of languages
  • Minimal maintenance through easily replaceable cartridges, a long lasting UV lamp and timely service alerts
  • Data connection: Ethernet (RJ45)
Additional bench space can be saved by placing the Milli-Q® Reference system under your bench or by using the optional bracket to wall mount the unit. If fed from a reservoir, the system can be protected with a feed water conductivity cell.
Certifikace: The Milli-Q® Reference system is tested by an independent and accredited company for compliance with the CE directives related to safety and electromagnetic compatibility. It is built using components and practices recommended by UL and has been CUL marked. The Milli-Q® Reference system is delivered with a Certificate of Conformity ensuring that it has been built and tested fully assembled following Millipore Standard Operating Procedures and a Certificate of Calibration for the temperature and resistivity meters built in the system. The Milli-Q® Reference consumables are automatically delivered with a Certificate of Quality. Millipore’s manufacturing site is ISO 9001 v.2000 and ISO 140001 certified.
Informace pro objednávání: A wide range of end filters allows to adapt the ultrapure water to a specific application in a glimpse: The user should switch from a Millipak® Express 40 filter for general laboratory applications to an LC-Pak for production of UHPLC or LC-MS grade water or a BioPak® filter for the delivery of pyrogen-free and nuclease-free water for PCR or other molecular biology applications.