Order Entry
VWR®, Density Standards for the Petroleum Industry, Certified Reference Materials
Supplier:  VWR Chemicals
Certificates
undefined
VWR®, Density Standards for the Petroleum Industry, Certified Reference Materials
Supplier:  VWR Chemicals
Certificates
Specifications
  • Environmentally Preferable:
Specifications
Related Information
Product Information
  • Catalog #
    Seq
    Density
    Reference temperature
    Pack type
    Pk
    Availability
    Price
  • Density standard, gasoline
    Density standard, gasoline
    1
    0,73632 g/ml (nominal)
    15 °C
    Aluminium bottle
    250 mL
    Log in for availability
    1 * 250 mL
    1 * 250 mL - 292,00 €
    292,00 €
    New Customer Promotional Price
    1 * 250 mL
       292,00 €
  • Density standard, jet aviation fuel
    Density standard, jet aviation fuel
    99110.230
    2
    0,79672 g/ml (nominal)
    15 °C
    Aluminium bottle
    250 mL
    Log in for availability
    1 * 250 mL
    1 * 250 mL - 292,00 €
    292,00 €
    New Customer Promotional Price
    1 * 250 mL
       292,00 €
  • Density standard, diesel
    Density standard, diesel
    3
    0,84238 g/ml (nominal)
    15 °C
    Aluminium bottle
    250 mL
    Log in for availability
    1 * 250 mL
    1 * 250 mL - 292,00 €
    292,00 €
    New Customer Promotional Price
    1 * 250 mL
       292,00 €
  • Density standard, lubricant
    Density standard, lubricant
    4
    0,86709 g/ml (nominal)
    15 °C
    Aluminium bottle
    250 mL
    Log in for availability
    1 * 250 mL
    1 * 250 mL - 292,00 €
    292,00 €
    New Customer Promotional Price
    1 * 250 mL
       292,00 €