Order Entry
VWR® INCU-Line®, Cooled Incubators
VWR® INCU-Line®, Cooled Incubators
undefined
VWR® INCU-Line®, Cooled Incubators
  • Catalog No:
  • 390-1341
  • 390-1342
  • Model:
  • IL 68R
  • IL 150R
  • Temperature fluctuation (time):
  • ≤±0,1 °C @ 37 °C
  • ≤±0,2 °C @ 37 °C
  • Temperature variation (spatial):
  • ≤±0,3 °C @ 37 °C
  • ≤±0,8 °C @ 37 °C
  • Temperature range:
  • 3…70 °C
  • 3…70 °C
  • Capacity:
  • 68 L
  • 143 L
  • Max. chamber workload:
  • 20 kg
  • 30 kg
  • Convection type:
  • Compressor cooling and heating elements
  • Compressor cooling and heating elements
  • Nominal power consumption:
  • 900 W
  • 1300 W
  • Weight:
  • 41 kg
  • 68 kg
  • Shelves supplied (max.):
  • 2 (2)
  • 3 (4)
  • Max. load per shelf:
  • 10 kg
  • 10 kg
  • Int. W×D×H:
  • 470×300×430 mm
  • 520×410×660 mm
  • Ext. W×D×H:
  • 570×680×660 mm
  • 620×650×920 mm
  •  
    Seq
    Type
    Capacity
    Pk
  • 390-1341
    001
    INCU-Line® IL 68R cooled incubator (EU-plug only)
    68 L
    1
    1 * 1 items
    1 * 1 items - 2 660,00 €
    2 660,00 €
    1 * 1 items
       2 660,00 €
    Accessories
    Specifications
    Model
    IL 68R
    Environmentally Preferable
    Temperature fluctuation (time)
    ≤±0,1 °C @ 37 °C
    Temperature variation (spatial)
    ≤±0,3 °C @ 37 °C
    Temperature range
    3…70 °C
    Capacity
    68 L
    Max. chamber workload
    20 kg
    Convection type
    Compressor cooling and heating elements
    Nominal power consumption
    900 W
    Weight
    41 kg
    Shelves supplied (max.)
    2 (2)
    Max. load per shelf
    10 kg
    Int. W×D×H
    470×300×430 mm
    Ext. W×D×H
    570×680×660 mm
  • 390-1342
    002
    INCU-Line® IL 150R cooled incubator (EU-plug only)
    143 L
    1
    1 * 1 items
    1 * 1 items - 3 160,00 €
    3 160,00 €
    1 * 1 items
       3 160,00 €
    Accessories
    Specifications
    Model
    IL 150R
    Environmentally Preferable
    Temperature fluctuation (time)
    ≤±0,2 °C @ 37 °C
    Temperature variation (spatial)
    ≤±0,8 °C @ 37 °C
    Temperature range
    3…70 °C
    Capacity
    143 L
    Max. chamber workload
    30 kg
    Convection type
    Compressor cooling and heating elements
    Nominal power consumption
    1300 W
    Weight
    68 kg
    Shelves supplied (max.)
    3 (4)
    Max. load per shelf
    10 kg
    Int. W×D×H
    520×410×660 mm
    Ext. W×D×H
    620×650×920 mm