Lead Free Tacky Flux, Circuitworks®
|
Danger
|
CircuitWorks® lead free tacky flux is a type ROL0 formulation engineered specifically for the higher temperatures required by lead free soldering of BGA rework that require high-reliability, stability and cleanliness.
- Excellent wetting ability
- Formulated for high temperature
- Long tack time
- Noncorrosive, halide and halogen free
- Excellent consistency
- Stable viscosity
CircuitWorks® lead free tacky flux gel composition holds the BGA component in position even with board movement. Its lower viscosity allows easy application and contains no ionic material. CircuitWorks® lead free tacky flux is suitable for clean room applications.
Certifikace: Conforms to ISO 9454. Meets Bellcore TR-NWT-000078 and DIN EN 29454-1 1.1.3.C classification.