Double bucket system, polypropylene
Supplier: Contec
129-0211EA 368 EUR
CONT2720EU
Double bucket system, polypropylene
Fully autoclavable PP rolling trolley with two 25 L buckets and wringer.
  • Durable plastic bucket system is lightweight and easy to maneuver
  • System includes two different colour buckets for easy solution identification
  • Fully autoclavable system is ideal for sterile environment use
Durable, yet lightweight construction makes Contec's 2720 bucket system easy to manoeuvre around cleanrooms. Red and blue buckets allow for differentiation between clean and waste solutions. Ideal for use with Contec's Edgeless® mop.
Delivery information: Includes two 25 L polypropylene buckets (red, blue), trolley, and wringer.