Drum wrench
119-0238
Specifications
- Description:Drum wrench
- Material:Brass
- Environmentally Preferable:
- Pk:1
Specifications
About this item
Drum Wrench opens drum bung caps.
- Rugged and non-sparking
- Design of handle protects the hand
This corrosive-resistant tool fits many 50,8 or 19,05 mm bung styles commonly used on drums.