Suspension trauma safety straps, DBI-SALA® Rollgliss™ RPD
Supplier: 3M
CPSD9501403EA 30.85 EUR
CPSD9501403
Suspension trauma safety straps, DBI-SALA® Rollgliss™ RPD
Suspension trauma safety straps, one pair, attaches to most harnesses. The strap is designed to help a worker overcome the potential negative health impacts of suspension trauma (otherwise known as orthostatic intolerance). The safety straps prevent suspension trauma while a worker is awaiting rescue. They also allow the worker to stand up in their harness and to relieve the pressure being applied to the arteries and veins around the top of the legs.
  • Continuous loop design relieves pressure on the legs
  • Fast and easy installation
  • Extremely compact and lightweight design
  • Simple and fast zipper style deployment
  • In-field re-setable design
The strap accommodates either having one foot or both feet in the loop at a time it will relieve the pressure to both sides with just one foot in allowing for added movement of the legs. The strap allows for increased comfort, balance and improved circulation in the legs while suspended and waiting for rescue.
Certifikace: EN 358