You Searched For: Adapters, Camera to Microscope
10  results were found
SearchResultCount:"10"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: These adapters enable cameras with different chip sizes to be connected to microscopes with a C-mount thread.
Supplier: VWR Collection
Description: C-mount adapter compatible with Nikon microscopes, 0,5×
Supplier: VWR Collection
Description: Eyepiece adapter, 0,45x, Ø: 23/30 mm
Supplier: VWR Collection
Description: Photo adapter for APS-C and Full Frame Reflex cameras
Supplier: VWR Collection
Description: C-mount adapter, focusable, 0,65× (for VWR fluorescence inverted microscope)
Catalog Number: (630-1747)
Supplier: VWR Collection
Description: Phototube adapter for digital camera
UOM: 1 * 1 items
Supplier: ZEISS
Description: Adapter, 60N - 44
Sale
Supplier: MOTIC
Description: CCD adapter for C-mount camera, 0,5×
Catalog Number: (IDCPMS30A)
Supplier: Dino-Lite / IDCP B.V.
Description: Adapter with standard thread to use the holder on third party tripods (not included)
UOM: 1 * 1 items
Supplier: EPPENDORF
Description: Microscope adapters for InjectMan® NI 2 and TransferMan® NK 2. The universal stand is suitable for mounting manipulators on upright or stereo microsco...
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 10 of 10
no targeter for Bottom