Boiler hydrometers
Supplier: Amarell
Certificates
About this item
Hydrometers with thermometer for boiler feed water according to Doctor Ammer.
- Shot weighted body
- Paper scale with well visible print
- Bottom pointed end
Product Information
- Catalog #Supplier #SeqRangeDivisionCalibration temperatureLengthPkAvailabilityPrice
- Boiler hydrometerBoiler hydrometerAMARH841660-FLH841660-FL011 - 0 - 10,1 %20 °C300 mm11 * 1 itemsSpecifications:
Specifications
Environmentally Preferable - Boiler hydrometerBoiler hydrometerAMARH841664-FLH841664-FL021 - 0 - 20,1 %20 °C300 mm11 * 1 itemsSpecifications:
Specifications
Environmentally Preferable - Boiler hydrometerBoiler hydrometerAMARH841668-FLH841668-FL030,5 - 0 - 0,50,01 %20 °C308 mm11 * 1 itemsSpecifications: