Shodex HILICpak VT-50 HILIC Columns for Anionic Analytes Including Glyphosate
Supplier: Resonac
Certificates
About this item
Robust polymer-based HILIC column for negatively charged analytes.
- Polymer-based HILIC columns with quaternary ammonium (-NR3+) groups
- Best for glyphosate analysis and phosphorylated sugars
- Suitable for anionic substances (especially phosphate compounds) analysis using HILIC mode
- Use of some eluents add ion exchange mode
- Polymer-based packing material provides excellent chemical stability and minimum deterioration over an extended time period
- Suitable for LC-MS/MS analysis
The HILICpak VT-50 column with quaternary ammonium groups is a robust HILIC column for anionic substance analysis. The polymeric base material polyvinyl alcohol offers great stability from pH 2 - 13. The column housing is made of PEEK to reduce unwanted interaction.
Specifications
- Catalog No:
- SHOWF6711300
- SHOWF7630400
- Description:
- HILICpak VT-50G 2A
- HILICpak VT-50 2D
- Max. pressure:
- 10 MPa (100 bar)
- pH range:
- 2 - 13
- 2 - 13
- Temperature limit:
- 4 - 60 °C
- 4 - 60 °C
Specifications
Product Information
- Catalog #Supplier #SeqColumn FormatPhaseColumn hardwarePore sizeParticle SizeAvailabilityPrice
- HILICpak VT-50G 2AHILICpak VT-50G 2ASHOWF6711300F671130001Guard columnQuaternary ammoniumPEEK5 µmLog in for availability1 * 1 items554,00 €Specifications:
More
Specifications
DescriptionHILICpak VT-50G 2AEnvironmentally PreferableMax. pressurepH range2 - 13Temperature limit4 - 60 °C - HILICpak VT-50 2DHILICpak VT-50 2DSHOWF7630400F763040002AnalyticalQuaternary ammoniumPEEK100 Å5 µmLog in for availability1 * 1 items1.770,00 €Specifications:
More
Specifications
DescriptionHILICpak VT-50 2DEnvironmentally PreferableMax. pressure10 MPa (100 bar)pH range2 - 13Temperature limit4 - 60 °C