ESD safety shoes, lace-up, L10, Breezer Black, Low, S1P
Supplier: Elten
ELTN721051/43EA 72.9 EUR
ELTN721051/43 ELTN721051/42 ELTN721051/41 ELTN721051/40 ELTN721051/47 ELTN721051/45 ELTN721051/44 ELTN721051/39 ELTN721051/38 ELTN721051/37
ESD safety shoes, lace-up, L10, Breezer Black, Low, S1P
Environmental friendly manufactured ESD safety shoe made from nubuck leather, a padded tongue and a low profile dual-density TPU/PU sole L10.
  • Breathable textile lining
  • Full-length inlay sole ESD PRO grey
  • Metal-free puncture resistant midsole
  • Protective steel toecap (200 J)
Ideal for use in dry work areas such as industry, storage, transport, or assembly and where there is a risk of penetration from pointed and sharp objects.
Colour: Black/grey
Certifikace: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S1P, SRC