Certificates
Chemical Structure
Manufacturer Data
Login
/
Register
Slovenia
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
0
0,00 €
Order Entry
2-Methylhydroquinone
:
Roth Carl
2-Methylhydroquinone
:
Roth Carl
:
Methylhydroquinone
:
95-71-6
:
:
C₇H₈O₂
:
MFCD00002345
:
285
:
124.14 g/mol
:
123…126 °C
:
Ambient
:
202-443-7
:
2811
:
6.1 III
:
GHS07
Environmentally Preferable:
Pack type
Pk
ROTH4787.1
4787.1
Glass bottle for solids
100 g
1 * 100 g
-
+
1 * 100 g
-
+
ROTH4787.2
4787.2
Glass bottle for solids
500 g
1 * 500 g
-
+
1 * 500 g
-
+