ESD safety shoes, sandals, uvex motion style, 6975
Supplier: Uvex
UVEX6975.2/40EA 108 EUR
UVEX6975.2/40 UVEX6975.2/45 UVEX6975.2/42
ESD safety shoes, sandals, uvex motion style, 6975
Ultralight ESD safety shoe made from breathable high-tech microvelour, soft padded collar and tongue, breathable distance mesh and an extremely flexible dual density PUR sole (slip-, abrasion- and cut-resistant, resistant to oil and petrol and withstands brief exposure to heat of approx. +120 °C). Free of lacquer wetting inhibitors such as silicones or phthalates.
  • Climazone technology for improved comfort
  • Breathable distance mesh lining
  • Anti-twist heel cap for better stability and protection from ankle twisting
  • Removable anti-static comfort insole with moisture control system; orthopaedic insoles possible
  • 3D hydroflex® foam footbed for optimal cushioning in the heel and forefoot area
  • Virtually seamless design to eliminate the potential for pressure points and blisters
  • Protective steel toecap (200 J)
  • Metal-free puncture resistant midsole
  • Fulfils ESD requirements: Resistance to ground inferior to 35 megaohms
Suitable for people allergic to chrome.
Colour: Black/silver
Width: 11
Certifikace: EN 61340 (ESD); EN ISO 20345:2011; S1 (6975.8), S1P (6975.2), SRC