Order Entry
Disposal Bins for Solvent Wastes
Disposal Bins for Solvent Wastes
 :  asecos
undefined
Disposal Bins for Solvent Wastes
 :  asecos
  •  
    Seq
    Colour
    Material
    Capacity
    W×D×H
    Pk
  • Disposal bins, round with foot pedal
    Disposal bins, round with foot pedal
    MSGA23156
    23156
    01
    Red
    Sheet metal
    35 L
    450×440×490 mm
    1
    1 * 1 items
    1 * 1 items - 178,50 €
    178,50 €
    1 * 1 items
       178,50 €
    Specifications
    Environmentally Preferable
  • Disposal bins, round with foot pedal
    Disposal bins, round with foot pedal
    MSGA23157
    23157
    02
    Red
    Sheet metal
    50 L
    505×495×540 mm
    1
    1 * 1 items
    1 * 1 items - 223,50 €
    223,50 €
    1 * 1 items
       223,50 €
    Specifications
    Environmentally Preferable
  • Disposal bins, round with foot pedal
    Disposal bins, round with foot pedal
    MSGA23158
    23158
    03
    Red
    Sheet metal
    65 L
    545×535×580 mm
    1
    1 * 1 items
    1 * 1 items - 266,00 €
    266,00 €
    1 * 1 items
       266,00 €
    Specifications
    Environmentally Preferable
  • Disposal bins, round with foot pedal
    Disposal bins, round with foot pedal
    MSGA23159
    23159
    04
    Stainless steel
    Stainless steel 1.4301
    35 L
    450×440×490 mm
    1
    1 * 1 items
    1 * 1 items - 272,00 €
    272,00 €
    1 * 1 items
       272,00 €
    Specifications
    Environmentally Preferable
  • Disposal bins, round with foot pedal
    Disposal bins, round with foot pedal
    MSGA23160
    23160
    05
    Stainless steel
    Stainless steel 1.4301
    50 L
    505×495×540 mm
    1
    1 * 1 items
    1 * 1 items - 287,50 €
    287,50 €
    1 * 1 items
       287,50 €
    Specifications
    Environmentally Preferable
  • Disposal bins, round with foot pedal
    Disposal bins, round with foot pedal
    MSGA23161
    23161
    06
    Stainless steel
    Stainless steel 1.4301
    65 L
    545×535×580 mm
    1
    1 * 1 items
    1 * 1 items - 334,50 €
    334,50 €
    1 * 1 items
       334,50 €