Wash gloves, SafeTouch® Medigloves Premium
HOPE102603EA 67.7 EUR
HOPE102603
Wash gloves, SafeTouch® Medigloves Premium
This ready-to-use bathing system eliminates the need to use bowls of water and towels. It provides effective patient care and infection control, prevents cross-contamination.
  • Impregnated with Aloe Vera and vitamin E to help to soothe, moisturise and protect the delicate skin
  • Non linting, spunlaced 3D structured wash gloves provide exceptionally soft and gentle bathing procedure
  • Cost effective – reduces the labor and material costs
  • Disposable, so no need to clean up after bath
  • Less physical strain for nurses – no need to lift, carry heavy bowls of water
  • Microwaveable
Ideal for nursing staff in intensive care wards and pain treatment where traditional washing is burdensome.
Balení: Re-sealable pack with eight wash gloves.