You Searched For: Glass and Cotton Wool
15  results were found
SearchResultCount:"15"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Chemicals
Description: Glass wool is often used as filter material in analytical processes and as supporting substance for drying agents in drying towers (e.g. to prevent “c...
Catalog Number: (734-2671)
Supplier: VWR Collection
Description: Thorough machine processing provides consistent uniformity of shape and size for maximum efficacy in vial plugging. These high quality balls are very ...
UOM: 1 * 2.000 items
Catalog Number: (24950.152)
Supplier: VWR Chemicals
Description: Our silica wool is made from pure fused quartz fiber: amorphous, continuous, tangled, white, odorless, with no volatile component and no shot. Length,...
UOM: 1 * 50 g
Supplier: TECHNICAL QUARTZ SOLUTIONS GMB
Description: Quartz wool, Fiberglass, White, 1000 g
Supplier: Thermo Scientific
Description: This glass wool can be used as a filter aid.
Supplier: GLASWARENFABRIK KARL HECHT
Description: Extra fine.
Catalog Number: (HYPE60180-785)
Supplier: Thermo Fisher Scientific
Description: Deactivated glass wool, 10 g, Fiberglass, White, 10 g
UOM: 1 * 1 items
Supplier: PanReac AppliChem
Description: Glass wool, washed, technical grade, Fiberglass, 250 g
Supplier: MACHEREY-NAGEL
Description: This glass wool can be used for GC liners, packed GC columns etc.
Supplier: MACHEREY-NAGEL
Description: Filter aids transform difficult precipitates and colloidal particles into a form which can be easily filtered. When slimy and strongly lyophilic, swel...
Supplier: Restek
Description: Deactivated borosilicate glass wool, more flexible than fused silica wool, Fiberglass, White, 10 g
Supplier: Agilent
Description: Glass wool, silane-treated, pesticide grade, for gas chromatography, 10 g, Fiberglass, White, 10 g
Catalog Number: (QUTW1186)
Supplier: QUARTZTECHNISCHE WERKSTATTEN
Description: Loosely, fibre size 2 to12 μm.
UOM: 1 * 500 g
Catalog Number: (BELC7750-22222)
Supplier: BELLCO GLASS INC.
Description: Cotton roving made of 12 grain premium long fiber cotton.
UOM: 1 * 1 kg
Supplier: Roth Carl
Description: This quartz wool is ideal for elementary analysis, as a final stopper in chromatography columns or as a filter aid.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 15 of 15
no targeter for Bottom