You Searched For: Ink Pens
8  results were found
SearchResultCount:"8"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: OUTILS RUBIS SA
Description: Hexagonal aluminium scribing pen, 8 mm, with replaceable diamond tip.
Catalog Number: (HECH41182010)
Supplier: GLASWARENFABRIK KARL HECHT
Description: Diamond pen, Application: Glass
UOM: 1 * 1 items
Supplier: USBECK Laborgeräte
Description: Diamond tip in metal holder.
Catalog Number: (MARI6120300)
Supplier: MARIENFELD
Description: For permanent labelling of microscope slides and other glass products. Indelibly engraved markings comply particularly with the guidelines of GLP.
UOM: 1 * 1 items
Supplier: Bel-Art Products, a Part of SP
Description: Heavy-duty soft tip disposable pen cornes in several colours and write on glass, metal, ceramics, plastics, paper etc. The oil-based marking paint res...
Supplier: MENSCH, FRANZ
Description: Hygostar® detectable ballpoint pens are available in blue, black and red. The colour of the case corresponds to the ink colour.
Catalog Number: (HAMMHSB780-01)
Supplier: HAMMACHER KARL
Description: Diamond for writing on glass.
UOM: 1 * 1 items
Catalog Number: (817-1135)
Supplier: Bel-Art Products, a Part of SP
Description: It can be used to correct or write on wet labels that were kept in a water bath, in liquid nitrogen or in a freezer at 0 °C. Can be also used on dry p...
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 8 of 8
no targeter for Bottom