You Searched For: Cleanroom Pens
7  results were found
SearchResultCount:"7"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: KEYSTONE CLEANROOM PRODUCTS
Description: These pens and markers eliminate the risks associated with surface sanitisation procedures.
Sale
Supplier: KEYSTONE CLEANROOM PRODUCTS
Description: These pens and markers eliminate the risks associated with surface sanitisation procedures.
Sale
Supplier: MICRONOVA
Description: Micronova cleanroom pens feature low sodium, indelible ink and a special low particulate barrel to limit particle generation. Cleanroom irradiated Sha...
Supplier: MICRONOVA
Description: A permanent marker that gives the thinnest permanent line at 0,3 mm.
Supplier: MICRONOVA
Description: The industry standard, original pen style, fine point, permanent marker.
Catalog Number: (NITRS-BPBP-1-BL_U)
Supplier: Nitritex
Description: BioCleanâ„¢ Permaflow cleanroom pens have fade and water-resistant fast drying permanent ink and feature an alcohol-resistant label for easy cleaning wi...
UOM: 1 * 3 items
Blocked
Supplier: VWR Collection
Description: Manufactured with very low sodium to prevent sodium contamination.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 7 of 7
no targeter for Bottom