These block heaters are multi-purpose units that are ideal for incubation and activation of cultures, enzyme reactions, blood urea nitrogen determinations, immunoassays, melting/boiling points and a variety of other laboratory procedures.
- Exceptional temperature uniformity and accuracy
- Accommodate interchangeable, modular heating blocks
These high-wattage, constant-temperature block heaters are economical and versatile. Each unit requires minimum bench space and features convenient front-mounted temperature adjustment knobs providing high and low temperature control. Each model accepts separate interchangeable modular blocks, which accommodate various sample enclosures such as vials, microplates, PCR strips, and a wide variety of tubes. And each block contains a thermometer well, and features close tube-and-block contact for maximum heat retention.
Units offer two separate temperature adjustment knobs providing reproducible low temperature and high temperature control. Low range knob adjusts from ambient to 100 °C and high range knob adjusts from 75 to 150 °C.
Certifikace: RoHS, UL and C-UL listed.
Informace pro objednávání: Heaters require a VWR modular heating block for operation that has to be ordered separately. Additional 230 V, 50/60 Hz models are also available. All units are supplied with detachable, three-wire cord (234 cm) and plug.
Upozornení: To avoid possible electrical hazard, do not fill well or block opening with water or other fluids. Unit is designed as a dry bath/incubator.