Human serum, gastric cancer (GC)
HCMSPB007C-1-GCEA 0 EUR
HCMSPB007C-1-GC
Human serum, gastric cancer (GC)
Human serum used for researching the causes, diagnosis, supportive care and treatment of gastric cancer (GC). Collected from IRB-consented donors diagnosed with GC.
No pre-screening is done on fresh products, but donors are tested on day of collection for Hepatitis B, Hepatitis C, and HIV 1/2, HTLV-I/II, HIV-1/HCV/HBV nucleic acid, WNV nucleic acid, Trypanasoma cruzi and Syphilis.
Storage: Store at −80 °C.
Upozornení: Intended for research use only and not for direct therapeutic or diagnostic use in human or animals, or for further manufacturing of therapeutic products or devices.