Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
    • In Stock (41)
  • Supplier
  • Refine by Suppliers
    Sort by:
Protein Purification Media
SearchPresentationType-VERTICAL
SearchResultCount:"184"
  List View Easy View (new)
Sort by:
Supplier:  GE Healthcare - Life Sciences
Description:   Phenyl Sepharose™ 6 Fast Flow is a well established, standard aromatic hydrophobic interaction chromatography (HIC) medium for capture and intermediat...
Supplier:  GE Healthcare - Life Sciences
Description:   Octyl Sepharose™ 4 Fast Flow resin is based on cross-linked 4% agarose. The aliphatic ligand is immobilised to the base matrix with an ether linkage. ...
UOM:  1 * 200 ml
Supplier:  GE Healthcare - Life Sciences
Description:   SOURCE™ 15RPC is designed for reversed phase chromatography (RPC) with full pH range stability and suitable for polishing steps in industrial processe...
UOM:  1 * 10 ml
Supplier:  GE Healthcare - Life Sciences
Description:   GammaBind™ Plus Sepharose™ is GammaBind G, Type 3, covalently immobilised to Sepharose™ CL-6B by malimide linkage. It is used for lab-scale affinity p...
Supplier:  GE Healthcare - Life Sciences
Description:   Excellent for high-performance purification of GST-tagged proteins.
Catalog Number: (17-6002-35)
Supplier:  GE Healthcare - Life Sciences
Description:   Immunoprecipitation is a highly specific and effective technique for analytical separations of target antigens from crude cell lysates. In combination...
UOM:  1 * 2 items
Supplier:  Pall Laboratory
Description:   Q, DEAE, and CM Ceramic HyperD™ F ion exchangers are high capacity sorbents specially designed for the efficient and scaleable purification of biomole...
Supplier:  BioVision
Description:   High binding capacity beads for affinity purification of GST tagged proteins.
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 8  of 184
  1  2  3  4  5  6  7  8  9  10  11  12  13  14  15  Next