HyClone™ Cell Boost™ 7a and 7b Feed Supplements, HyClone products (Cytiva)
Supplier: HyClone products (Cytiva)
HyClone™
SH31026.01EA 1038 EUR
SH31026.01 SH31026.07 SH31027.07 SH31027.04
HyClone™ Cell Boost™ 7a and 7b Feed Supplements, HyClone products (Cytiva)
Cell Culture Supplements and Additives
Cell Boost™ 7a and 7b are feed additives especially matched to ActiPro™ production medium. Combining Cell Boost™ 7a and 7b has demonstrated significantly increased productivity when compared with batch culture.
These supplements are used as an essential part of a cell culture feed strategy to enhance cell culture performance and increase product yield. Cell Boost 7a and 7b are intended for use together in defined concentrations. Established protocols facilitate easy and efficient identification of a feeding strategy. Cell Boost 7a contains concentrated amino acids, vitamins, salts, trace elements, and a carbon source. Cell Boost 7b is a balanced formulation of selected amino acids in a concentrated form.
Informace pro objednávání: Lead times and minimum order quantities apply. Please contact your local Avantor sales office for more information.