Rinse aid, concentrate, neodisher® TS
Supplier: Dr. Weigert
neodisher®
WEIG310926EA 0 EUR
WEIG310926 WEIG310903
Rinse aid, concentrate, neodisher® TS
Cleaning Agents Detergents and Surface Cleansers
For rinsing dishes, food containers and trolleys in washers in all food processing companies, such as professional kitchens, butcher’s shops and bakeries.
  • Reliably dries even difficult-to-wet materials, such as e.g. plastic
  • Highly concentrated
  • Acidic formula, neutralises carried-over alkali residues and alkaline water
Also suitable for rinsing animal cages, accessories and racks for cages, storage and transport in specially designed washers.