These two-channel radio probes are equipped with an internal humidity and temperature probe, which makes them ideal for monitoring warehouse goods that are sensitive to temperature and humidity.
- Probe also monitors air conditioning in buildings
- Reliable measurement results based on latest measuring technology
- Integrated sensors guarantee readings with long-term stability
- With a data memory of 6 000 readings per channel
- Features a flexible wireless data transfer
H3 D radio probe equipped with a display, showing the current measurement data, battery status, limit value violations and quality of the radio link.
Weight: 245 g
* At 25 °C
Informace pro objednávání: Unit includes a wall bracket, calibration protocol, and 4×AA alkaline manganese batteries. Saveris converter V 2.0 (must be ordered separately) is required for integrating Saveris radio probes into systems with base firmware V 1.X.