Temperature/humidity radio probes, two-channel, Saveris H3 and H3 D
Supplier: Testo
TEST0572.6220
Temperature/humidity radio probes, two-channel, Saveris H3 and H3 D
These two-channel radio probes are equipped with an internal humidity and temperature probe, which makes them ideal for monitoring warehouse goods that are sensitive to temperature and humidity.
  • Probe also monitors air conditioning in buildings
  • Reliable measurement results based on latest measuring technology
  • Integrated sensors guarantee readings with long-term stability
  • With a data memory of 6 000 readings per channel
  • Features a flexible wireless data transfer
H3 D radio probe equipped with a display, showing the current measurement data, battery status, limit value violations and quality of the radio link.
Weight: 245 g
* At 25 °C
Informace pro objednávání: Unit includes a wall bracket, calibration protocol, and 4×AA alkaline manganese batteries. Saveris converter V 2.0 (must be ordered separately) is required for integrating Saveris radio probes into systems with base firmware V 1.X.